Imprimer

Anti-LMX1B Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314159EA 564 EUR
ORIGTA314159
Anti-LMX1B Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: LMX1B (LIM homeobox transcription factor 1, beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR