Anti-SCGB2A1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314191EA 564 EUR
ORIGTA314191
Anti-SCGB2A1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: SCGB2A1 (secretoglobin, family 2A, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR