Anti-MGST1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314192EA 564 EUR
ORIGTA314192
Anti-MGST1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: MGST1 (microsomal glutathione S-transferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR