Imprimer

Anti-MGST3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314194EA 543 EUR
ORIGTA314194
Anti-MGST3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: MGST3 (microsomal glutathione S-transferase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR