Anti-MAP3K10 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314202EA 564 EUR
ORIGTA314202
Anti-MAP3K10 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: MAP3K10 (mitogen-activated protein kinase kinase kinase 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR