Anti-MMP8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314204EA 552 EUR
ORIGTA314204
Anti-MMP8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: MMP8 (matrix metallopeptidase 8 (neutrophil collagenase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR