Anti-MOS Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314212EA 532 EUR
ORIGTA314212
Anti-MOS Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000 IELISA: 1:5000

Type: Primary
Antigen: MOS (v-mos Moloney murine sarcoma viral oncogene homolog)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR