Anti-NFKBIE Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314256EA 543 EUR
ORIGTA314256
Anti-NFKBIE Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: NFKBIE (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR