Anti-NOS2 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314267
ORIGTA314267EA 564 EUR
ORIGTA314267
Anti-NOS2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR