Imprimer

Anti-ROR2 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314292EA 564 EUR
ORIGTA314292
Anti-ROR2 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ROR2 (receptor tyrosine kinase-like orphan receptor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR