Anti-SLC22A18 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314311
ORIGTA314311EA 528 EUR
ORIGTA314311
Anti-SLC22A18 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: SLC22A18 (solute carrier family 22, member 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR