Anti-PIGH Rabbit Polyclonal Antibody

Supplier: OriGene
TA314340
ORIGTA314340EA 543 EUR
ORIGTA314340
Anti-PIGH Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: PIGH (phosphatidylinositol glycan anchor biosynthesis, class H)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR