Anti-PLCB3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314347EA 532 EUR
ORIGTA314347
Anti-PLCB3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 500-1:1000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: PLCB3 (phospholipase C, beta 3 (phosphatidylinositol-specific))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR