Anti-PPP1R2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314369EA 564 EUR
ORIGTA314369
Anti-PPP1R2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions:

Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR