Anti-PKIB Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314384EA 543 EUR
ORIGTA314384
Anti-PKIB Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: PKIB (protein kinase (cAMP-dependent, catalytic) inhibitor beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR