Imprimer

Anti-PSMD11 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314396EA 564 EUR
ORIGTA314396
Anti-PSMD11 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: PSMD11 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR