Anti-PTGDR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314400EA 564 EUR
ORIGTA314400
Anti-PTGDR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: PTGDR (prostaglandin D2 receptor (DP))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR