Anti-RPS21 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314495
ORIGTA314495EA 528 EUR
ORIGTA314495
Anti-RPS21 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: RPS21 (ribosomal protein S21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR