Imprimer

Anti-SCN7A Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314512EA 564 EUR
ORIGTA314512
Anti-SCN7A Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: SCN7A (sodium channel, voltage-gated, type VII, alpha subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR