Anti-SCNN1D Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314516EA 564 EUR
ORIGTA314516
Anti-SCNN1D Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: SCNN1D (sodium channel, non-voltage-gated 1, delta subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR