Anti-SFSWAP Rabbit Polyclonal Antibody

Supplier: OriGene
TA314525
ORIGTA314525EA 528 EUR
ORIGTA314525
Anti-SFSWAP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000 References:

Type: Primary
Antigen: SFSWAP (splicing factor, suppressor of white-apricot homolog (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR