Anti-SOX12 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314556
ORIGTA314556EA 528 EUR
ORIGTA314556
Anti-SOX12 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: SOX12 (SRY (sex determining region Y)-box 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR