Imprimer

Anti-SS18 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314572EA 564 EUR
ORIGTA314572
Anti-SS18 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: SS18 (synovial sarcoma translocation, chromosome 18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR