Imprimer

Anti-TEAD1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314607EA 552 EUR
ORIGTA314607
Anti-TEAD1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: TEAD1 (TEA domain family member 1 (SV40 transcriptional enhancer factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR