Anti-TNFAIP2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314633EA 564 EUR
ORIGTA314633
Anti-TNFAIP2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: TNFAIP2 (tumor necrosis factor, alpha-induced protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR