Imprimer

Anti-TOP2B Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314637EA 564 EUR
ORIGTA314637
Anti-TOP2B Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: TOP2B (topoisomerase (DNA) II beta 180kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR