Anti-YWHAH Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314676EA 543 EUR
ORIGTA314676
Anti-YWHAH Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: YWHAH (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR