Imprimer

Anti-YWHAZ Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314679EA 564 EUR
ORIGTA314679
Anti-YWHAZ Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR