Imprimer

Anti-DGKD Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314762EA 564 EUR
ORIGTA314762
Anti-DGKD Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: DGKD (diacylglycerol kinase, delta 130kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR