Anti-EIF3F Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314786EA 552 EUR
ORIGTA314786
Anti-EIF3F Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: EIF3F (eukaryotic translation initiation factor 3, subunit F)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR