Imprimer

Anti-PEA15 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314790EA 564 EUR
ORIGTA314790
Anti-PEA15 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: PEA15 (phosphoprotein enriched in astrocytes 15)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR