Imprimer

Anti-SERPINB7 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314799EA 564 EUR
ORIGTA314799
Anti-SERPINB7 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: SERPINB7 (serpin peptidase inhibitor, clade B (ovalbumin), member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR