Imprimer

Anti-MYOM1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314805EA 543 EUR
ORIGTA314805
Anti-MYOM1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: MYOM1 (myomesin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR