Anti-TNFSF12 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314806EA 552 EUR
ORIGTA314806
Anti-TNFSF12 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: TNFSF12 (tumor necrosis factor (ligand) superfamily, member 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR