Imprimer

Anti-TRIM24 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314810EA 564 EUR
ORIGTA314810
Anti-TRIM24 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: TRIM24 (tripartite motif containing 24)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR