Anti-EIF2B4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314830EA 564 EUR
ORIGTA314830
Anti-EIF2B4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: EIF2B4 (eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR