Anti-MED21 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314905
ORIGTA314905EA 564 EUR
ORIGTA314905
Anti-MED21 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: MED21 (mediator complex subunit 21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR