Anti-C14orf2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314933EA 564 EUR
ORIGTA314933
Anti-C14orf2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: C14orf2 (chromosome 14 open reading frame 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR