Anti-ARHGAP11A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA314972EA 543 EUR
ORIGTA314972
Anti-ARHGAP11A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: ARHGAP11A (Rho GTPase activating protein 11A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR