Imprimer

Anti-SEC16A Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314989EA 564 EUR
ORIGTA314989
Anti-SEC16A Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: SEC16A (SEC16 homolog A (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR