Imprimer

Anti-ZBTB40 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA314990EA 543 EUR
ORIGTA314990
Anti-ZBTB40 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: ZBTB40 (zinc finger and BTB domain containing 40)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR