Anti-NR1I3 Rabbit Polyclonal Antibody

Supplier: OriGene
TA314998
ORIGTA314998EA 564 EUR
ORIGTA314998
Anti-NR1I3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: NR1I3 (nuclear receptor subfamily 1, group I, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR