Imprimer

Anti-COX17 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315013EA 564 EUR
ORIGTA315013
Anti-COX17 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: COX17 (COX17 cytochrome c oxidase copper chaperone)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR