Imprimer

Anti-SF3B4 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315039EA 564 EUR
ORIGTA315039
Anti-SF3B4 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: SF3B4 (splicing factor 3b, subunit 4, 49kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR