Anti-SLC30A9 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315056EA 564 EUR
ORIGTA315056
Anti-SLC30A9 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: SLC30A9 (solute carrier family 30 (zinc transporter), member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR