Imprimer

Anti-ATP5H Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA315059EA 543 EUR
ORIGTA315059
Anti-ATP5H Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: ATP5H (ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR