Anti-BATF Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315071EA 564 EUR
ORIGTA315071
Anti-BATF Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:1000

Type: Primary
Antigen: BATF (basic leucine zipper transcription factor, ATF-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR