Anti-ARFGEF2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315073EA 564 EUR
ORIGTA315073
Anti-ARFGEF2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: ARFGEF2 (ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR