Anti-POLQ Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315096EA 545 EUR
ORIGTA315096
Anti-POLQ Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: POLQ (polymerase (DNA directed), theta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR