Anti-TCFL5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA315099EA 564 EUR
ORIGTA315099
Anti-TCFL5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: TCFL5 (transcription factor-like 5 (basic helix-loop-helix))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR